Assay Details
Target Gene Details
Entrez Gene ID: | 23640 |
Gene Name: | HSPA (Hsp70) binding protein 1 |
Gene Aliases: |
FES1 |
Location: |
Chr.19:55262222-55280383 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPBP1 | NM_001130106.1 | NP_001123578.1 | ||
| NM_001297600.1 | NP_001284529.1 | |||
| NM_012267.4 | NP_036399.3 | |||
| XM_005258700.2 | XP_005258757.1 | |||
| XM_005258701.2 | XP_005258758.1 | |||
| XM_011526691.1 | XP_011524993.1 | |||
| XM_017026543.1 | XP_016882032.1 | |||
| XM_017026544.1 | XP_016882033.1 | |||
| XM_017026545.1 | XP_016882034.1 | |||
| AB020592.1 | BAB18742.1 | |||
| AF093420.1 | AAC79703.1 | |||
| AF187859.1 | AAF35833.1 | |||
| AF217996.1 | AAG17238.1 | |||
| AK075293.1 | ||||
| AK129755.1 | ||||
| AK130781.1 | ||||
| AK294358.1 | ||||
| BC001236.1 | AAH01236.1 | |||
| BC002373.2 | AAH02373.1 | |||
| CR457118.1 | CAG33399.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs67113555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map