Assay Details
Target Gene Details
Entrez Gene ID: | 10055 |
Gene Name: | SUMO1 activating enzyme subunit 1 |
Gene Aliases: |
AOS1, HSPC140, SUA1, UBLE1A |
Location: |
Chr.19:47130782-47210636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAE1 | NM_001145713.1 | NP_001139185.1 | ||
| NM_001145714.1 | NP_001139186.1 | |||
| NM_005500.2 | NP_005491.1 | |||
| NR_027280.1 | ||||
| XM_006722963.2 | XP_006723026.1 | |||
| XM_006722965.3 | XP_006723028.1 | |||
| XM_017026135.1 | XP_016881624.1 | |||
| XM_017026136.1 | XP_016881625.1 | |||
| AF046025.2 | AAD23902.2 | |||
| AF090385.2 | AAD12785.2 | |||
| AF110956.1 | AAD24433.1 | |||
| AF161489.1 | AAF29104.1 | |||
| AK021978.1 | ||||
| AK027595.1 | ||||
| AK055481.1 | ||||
| AK090645.1 | ||||
| AK302287.1 | ||||
| AK315624.1 | ||||
| BC000344.2 | AAH00344.1 | |||
| BC003611.2 | AAH03611.1 | |||
| BC018271.1 | AAH18271.1 | |||
| BT007290.1 | AAP35954.1 | |||
| DA047601.1 | ||||
| DA086973.1 | ||||
| DC297573.1 | ||||
| DC312737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833852 | Chr.19:47134590 - 47307834 on Build GRCh38 | Loss |
|
| nsv833851 | Chr.19:47058544 - 47208779 on Build GRCh38 | Loss |
|
| nsv1067417 | Chr.19:47103045 - 47309908 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150240456] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map