Assay Details
Target Gene Details
Entrez Gene ID: | 51477 |
Gene Name: | inositol-3-phosphate synthase 1 |
Gene Aliases: |
INO1, INOS, IPS, IPS 1, IPS-1 |
Location: |
Chr.19:18434388-18438301 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISYNA1 | NM_001170938.1 | NP_001164409.1 | ||
| NM_016368.4 | NP_057452.1 | |||
| NR_045573.1 | ||||
| NR_045574.1 | ||||
| XM_011528059.1 | XP_011526361.1 | |||
| AF086921.1 | AAP97151.1 | |||
| AF207640.1 | AAG35698.1 | |||
| AF220530.1 | AAF26444.1 | |||
| AK001325.1 | BAA91626.1 | |||
| AK021526.1 | BAB13837.1 | |||
| AK092179.1 | ||||
| AK093461.1 | ||||
| AK296022.1 | ||||
| BC018952.2 | AAH18952.1 | |||
| BC066902.1 | AAH66902.1 | |||
| DA615868.1 | ||||
| DB056699.1 | ||||
| DB244754.1 | ||||
| DC371373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677302 | Chr.19:18428535 - 18438882 on Build GRCh38 | Deletion |
|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv482217 | Chr.19:18434815 - 18438133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map