Assay Details
Target Gene Details
Entrez Gene ID: | 284415 |
Gene Name: | V-set and transmembrane domain containing 1 |
Gene Aliases: |
SIRL-1, SIRL1, UNQ3033 |
Location: |
Chr.19:54040825-54063966 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM1 | NM_001288791.1 | NP_001275720.1 | ||
| NM_001288792.1 | NP_001275721.1 | |||
| NM_001288793.1 | NP_001275722.1 | |||
| NM_198481.3 | NP_940883.2 | |||
| NR_110142.1 | ||||
| XM_011526845.1 | XP_011525147.1 | |||
| XM_011526846.1 | XP_011525148.1 | |||
| XM_011526847.1 | XP_011525149.1 | |||
| XM_011526848.1 | XP_011525150.1 | |||
| XM_011526849.1 | XP_011525151.1 | |||
| XM_017026666.1 | XP_016882155.1 | |||
| AY358542.1 | AAQ88906.1 | |||
| BC100943.2 | AAI00944.1 | |||
| BQ446114.1 | ||||
| DQ479397.1 | ||||
| FJ584316.1 | ||||
| FJ584317.1 | ||||
| FJ584318.1 | ||||
| FJ584319.1 | ||||
| FJ584320.1 | ||||
| FJ882051.1 | ||||
| FN398145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458782 | Chr.19:53961144 - 54093354 on Build GRCh38 | Gain |
|
| nsv510773 | Chr.19:54017180 - 54132236 on Build GRCh38 | Deletion |
|
| nsv1063127 | Chr.19:53926876 - 54068977 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv833877 | Chr.19:53951529 - 54117323 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72626273] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map