Assay Details
Target Gene Details
Entrez Gene ID: | 8425 |
Gene Name: | latent transforming growth factor beta binding protein 4 |
Gene Aliases: |
ARCL1C, LTBP-4, LTBP4L, LTBP4S |
Location: |
Chr.19:40592888-40629820 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTBP4 | XM_011527387.1 | XP_011525689.1 |
More Information
Set Membership: |
|

Genomic Map