Assay Details
Target Gene Details
Entrez Gene ID: | 84658 |
Gene Name: | adhesion G protein-coupled receptor E3 |
Gene Aliases: |
EMR3 |
Location: |
Chr.19:14600175-14674918 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE3 | AL832663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|

Genomic Map