Assay Details
Target Gene Details
Entrez Gene ID: | 3594 |
Gene Name: | interleukin 12 receptor subunit beta 1 |
Gene Aliases: |
CD212, IL-12R-BETA1, IL12RB, IMD30 |
Location: |
Chr.19:18058994-18099027 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470130 | Chr.19:17936474 - 18123777 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map