Assay Details
Target Gene Details
Entrez Gene ID: | 645432 |
Gene Name: | arrestin domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.19:4867614-4915469 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC5 | XM_017027144.1 | XP_016882633.1 | ||
| XM_017027145.1 | XP_016882634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762022 | Chr.19:4840987 - 4886538 on Build GRCh38 | Loss |
|
| esv3584953 | Chr.19:4844140 - 4886538 on Build GRCh38 | Gain |
|
| esv3643515 | Chr.19:4865553 - 4870303 on Build GRCh38 | Loss |
|
| nsv528462 | Chr.19:4852325 - 4878596 on Build GRCh38 | Loss |
|
| nsv470117 | Chr.19:4811659 - 4900672 on Build GRCh38 | Loss |
|
| nsv578455 | Chr.19:4851838 - 4878596 on Build GRCh38 | Loss |
|
| esv3893158 | Chr.19:4856682 - 4900672 on Build GRCh38 | Loss |
|
| nsv1065556 | Chr.19:4844193 - 4945238 on Build GRCh38 | Loss |
|
| esv2664700 | Chr.19:4864342 - 4871526 on Build GRCh38 | Deletion |
|

Genomic Map