Assay Details
Target Gene Details
Entrez Gene ID: | 593 |
Gene Name: | branched chain keto acid dehydrogenase E1, alpha polypeptide |
Gene Aliases: |
BCKDE1A, MSU, MSUD1, OVD1A |
Location: |
Chr.19:41397789-41425005 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCKDHA | NM_000709.3 | NP_000700.1 | ||
| NM_001164783.1 | NP_001158255.1 | |||
| AB209828.1 | BAD93065.1 | |||
| AK298188.1 | ||||
| AK313300.1 | ||||
| AL833143.1 | CAH56215.1 | |||
| BC007878.2 | AAH07878.1 | |||
| BC008933.2 | AAH08933.1 | |||
| BC023983.1 | AAH23983.1 | |||
| BI910860.1 | ||||
| J04474.1 | AAB59549.1 | |||
| Z14093.1 | CAA78475.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs368029947] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map