Assay Details
Target Gene Details
Entrez Gene ID: | 5819 |
Gene Name: | nectin cell adhesion molecule 2 |
Gene Aliases: |
CD112, HVEB, PRR2, PVRL2, PVRR2 |
Location: |
Chr.19:44846136-44889228 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECTIN2 | NM_001042724.1 | NP_001036189.1 | ||
| NM_002856.2 | NP_002847.1 | |||
| XM_011527192.2 | XP_011525494.1 | |||
| AF058448.1 | AAC23797.1 | |||
| AK291330.1 | ||||
| BC003091.1 | AAH03091.1 | |||
| CR456818.1 | CAG33099.1 | |||
| X80038.1 | CAA56342.1 |
More Information
Set Membership: |
|

Genomic Map