Assay Details
Target Gene Details
Entrez Gene ID: | 9622 |
Gene Name: | kallikrein related peptidase 4 |
Gene Aliases: |
AI2A1, ARM1, EMSP, EMSP1, KLK-L1, PRSS17, PSTS, kallikrein |
Location: |
Chr.19:50906352-50910745 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK4 | NM_001302961.1 | NP_001289890.1 | ||
| NM_004917.4 | NP_004908.4 | |||
| NR_126566.1 | ||||
| XM_011527545.2 | XP_011525847.1 | |||
| XM_011527546.2 | XP_011525848.1 | |||
| AF113140.1 | AAD21580.1 | |||
| AF259969.1 | AAF81227.1 | |||
| AY923170.1 | AAX30051.1 | |||
| BC069325.1 | AAH69325.1 | |||
| BC069403.1 | AAH69403.1 | |||
| BC069429.1 | AAH69429.1 | |||
| BC069489.1 | AAH69489.1 | |||
| BC096175.1 | AAH96175.1 | |||
| BC096176.3 | AAH96176.1 | |||
| BC096177.3 | ||||
| BC096178.3 | AAH96178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583473 | Chr.19:50906008 - 50922148 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map