Assay Details
Target Gene Details
Entrez Gene ID: | 284361 |
Gene Name: | ER membrane protein complex subunit 10 |
Gene Aliases: |
C19orf63, HSM1, HSS1 |
Location: |
Chr.19:50476477-50505905 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC10 | XM_011526818.2 | XP_011525120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map