Assay Details
Target Gene Details
Entrez Gene ID: | 53827 |
Gene Name: | FXYD domain containing ion transport regulator 5 |
Gene Aliases: |
DYSAD, HSPC113, IWU1, KCT1, OIT2, PRO6241, RIC |
Location: |
Chr.19:35154722-35169885 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXYD5 | NM_001164605.1 | NP_001158077.1 | ||
| NM_001320912.1 | NP_001307841.1 | |||
| NM_001320913.1 | NP_001307842.1 | |||
| NM_014164.5 | NP_054883.3 | |||
| NM_144779.2 | NP_659003.1 | |||
| NR_028406.1 | ||||
| AA044211.1 | ||||
| AB072911.1 | BAB83766.1 | |||
| AF161462.1 | AAF29077.1 | |||
| AK304012.1 | ||||
| AY157582.1 | AAO13166.1 | |||
| AY358991.1 | AAQ89350.1 | |||
| BC009642.2 | AAH09642.1 | |||
| BG025158.1 | ||||
| BM550889.1 | ||||
| BM839241.1 | ||||
| BQ183683.1 | ||||
| BQ440098.1 | ||||
| BQ941279.1 | ||||
| BT007343.1 | AAP36007.1 | |||
| BU164524.1 | ||||
| BU178888.1 |
More Information
Set Membership: |
|

Genomic Map