Assay Details
Target Gene Details
Entrez Gene ID: | 9454 |
Gene Name: | homer scaffolding protein 3 |
Gene Aliases: |
HOMER-3, VESL3 |
Location: |
Chr.19:18929201-18941258 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOMER3 | NM_004838.3 | NP_004829.3 | ||
| XM_006722943.1 | XP_006723006.1 | |||
| AL529052.3 | ||||
| BC012113.1 | AAH12113.1 |
Target Gene Details
Entrez Gene ID: | 102724360 |
Gene Name: | uncharacterized LOC102724360 |
Gene Aliases: |
- |
Location: |
Chr.19:18940254-18946831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724360 | NR_134912.1 | |||
| NR_134913.1 | ||||
| CF127232.1 | ||||
| DA366162.1 | ||||
| HY133933.1 | ||||
| HY154431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470132 | Chr.19:18853633 - 18943911 on Build GRCh38 | Loss |
|
| esv2667412 | Chr.19:18938136 - 18941983 on Build GRCh38 | Deletion |
|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv952423 | Chr.19:18859292 - 18941091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map