Assay Details
Target Gene Details
Entrez Gene ID: | 147945 |
Gene Name: | NLR family pyrin domain containing 4 |
Gene Aliases: |
CLR19.5, CT58, NALP4, PAN2, PYPAF4, RNH2 |
Location: |
Chr.19:55836578-55881855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP4 | NM_134444.4 | NP_604393.2 | ||
| AK315003.1 | ||||
| BC050326.1 | AAH50326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644849 | Chr.19:55836319 - 55989037 on Build GRCh38 | Gain |
|
| nsv580346 | Chr.19:55735154 - 55857948 on Build GRCh38 | Gain |
|
| esv3575075 | Chr.19:55820834 - 55846724 on Build GRCh38 | Gain |
|
| dgv6582n54 | Chr.19:55807428 - 56027874 on Build GRCh38 | Gain |
|
| esv3644850 | Chr.19:55838221 - 55841794 on Build GRCh38 | Loss |
|
| nsv1064105 | Chr.19:55808050 - 55939166 on Build GRCh38 | Gain |
|
| esv3893253 | Chr.19:55805430 - 55857497 on Build GRCh38 | Loss |
|
| nsv1057383 | Chr.19:55699665 - 55944662 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73622417] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map