Assay Details
Target Gene Details
Entrez Gene ID: | 6223 |
Gene Name: | ribosomal protein S19 |
Gene Aliases: |
DBA, DBA1, S19 |
Location: |
Chr.19:41859918-41871416 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS19 | NM_001022.3 | NP_001013.1 | ||
| NM_001321483.1 | NP_001308412.1 | |||
| NM_001321484.1 | NP_001308413.1 | |||
| NM_001321485.1 | NP_001308414.1 | |||
| AA165492.1 | ||||
| AK311786.1 | ||||
| BC000023.2 | AAH00023.1 | |||
| BC007615.2 | AAH07615.1 | |||
| BC017386.1 | AAH17386.1 | |||
| BC018616.1 | AAH18616.1 | |||
| BG398397.1 | ||||
| BU603942.1 | ||||
| CB124389.1 | ||||
| CB125125.1 | ||||
| M81757.1 | AAA89070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map