Assay Details
Target Gene Details
Entrez Gene ID: | 1089 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 4 |
Gene Aliases: |
CGM7, CGM7_HUMAN, NCA |
Location: |
Chr.19:41616986-41627087 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM4 | XM_011526348.2 | XP_011524650.1 | ||
| XM_017026200.1 | XP_016881689.1 | |||
| XM_017026203.1 | XP_016881692.1 | |||
| XM_017026208.1 | XP_016881697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718577 | Chr.19:41576456 - 41709018 on Build GRCh38 | Deletion |
|
| nsv519408 | Chr.19:41559909 - 41667978 on Build GRCh38 | Gain |
|
| nsv2494 | Chr.19:41612811 - 41648542 on Build GRCh38 | Insertion |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
| dgv3549n100 | Chr.19:41550966 - 41671232 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map