Assay Details
Target Gene Details
Entrez Gene ID: | 3383 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
BB2, CD54, P3.58 |
Location: |
Chr.19:10270841-10286615 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICAM1 | NM_000201.2 | NP_000192.2 | ||
| AF340038.1 | AAQ14901.1 | |||
| AF340039.1 | AAQ14902.1 | |||
| AK130659.1 | ||||
| AK298054.1 | ||||
| AK298983.1 | ||||
| AK301412.1 | ||||
| AK312636.1 | ||||
| BC015969.2 | AAH15969.1 | |||
| BP346334.1 | ||||
| BT006854.1 | AAP35500.1 | |||
| J03132.1 | AAA52709.1 | |||
| M24283.1 | AAA52708.1 | |||
| X06990.1 | CAA30051.1 |
More Information
Set Membership: |
|

Genomic Map