Assay Details
Target Gene Details
Entrez Gene ID: | 3609 |
Gene Name: | interleukin enhancer binding factor 3 |
Gene Aliases: |
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80 |
Location: |
Chr.19:10654261-10692419 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILF3 | NM_001137673.1 | NP_001131145.1 | ||
| NM_004516.3 | NP_004507.2 | |||
| NM_012218.3 | NP_036350.2 | |||
| NM_017620.2 | NP_060090.2 | |||
| NM_153464.2 | NP_703194.1 | |||
| XM_006722742.2 | XP_006722805.1 | |||
| XM_011527984.2 | XP_011526286.2 | |||
| XM_011527985.2 | XP_011526287.1 | |||
| XM_017026763.1 | XP_016882252.1 | |||
| XM_017026765.1 | XP_016882254.1 | |||
| AB209062.1 | BAD92299.1 | |||
| AF141870.1 | AAD37575.1 | |||
| AK291205.1 | ||||
| AK291617.1 | ||||
| AK293170.1 | ||||
| AK294084.1 | ||||
| BC048314.1 | AAH48314.1 | |||
| BC064836.1 | AAH64836.1 | |||
| DC320902.1 | ||||
| U10324.1 | AAA20994.1 | |||
| X98264.1 | CAA66917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map