Assay Details
Target Gene Details
Entrez Gene ID: | 4680 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 6 |
Gene Aliases: |
CD66c, CEAL, NCA |
Location: |
Chr.19:41755421-41772211 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM6 | NM_002483.6 | NP_002474.4 | ||
| XM_011526990.1 | XP_011525292.1 | |||
| AK312542.1 | ||||
| BC005008.1 | AAH05008.1 | |||
| BT009774.1 | AAP88776.1 | |||
| M18216.1 | AAA51739.1 | |||
| M18728.1 | AAA59907.1 | |||
| M29541.1 | AAA59915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2495 | Chr.19:41758303 - 41789908 on Build GRCh38 | Insertion |
|
| nsv579615 | Chr.19:41717515 - 41806347 on Build GRCh38 | Gain |
|
| esv3893209 | Chr.19:41752698 - 41776423 on Build GRCh38 | Loss |
|
| esv2763205 | Chr.19:41754639 - 41786690 on Build GRCh38 | Loss |
|
| nsv960844 | Chr.19:41760762 - 41765955 on Build GRCh38 | Duplication |
|
| esv3644394 | Chr.19:41749659 - 41789320 on Build GRCh38 | Gain |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map