Assay Details
Target Gene Details
Entrez Gene ID: | 729767 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 18 |
Gene Aliases: |
- |
Location: |
Chr.19:51478543-51491261 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM18 | NM_001278392.1 | NP_001265321.1 | ||
| XM_011527258.2 | XP_011525560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758767 | Chr.19:51488642 - 52101372 on Build GRCh38 | Gain+Loss |
|

Genomic Map