Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5SL | XM_011527065.2 | XP_011525367.1 |
Target Gene Details
Entrez Gene ID: | 100170765 |
Gene Name: | glutamate rich 4 |
Gene Aliases: |
C19orf69 |
Location: |
Chr.19:41442426-41444765 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH4 | XM_017026119.1 | 1 | 488 | XP_016881608.1 |
More Information
Set Membership: |
|

Genomic Map