Assay Details
Target Gene Details
Entrez Gene ID: | 6511 |
Gene Name: | solute carrier family 1 member 6 |
Gene Aliases: |
EAAT4 |
Location: |
Chr.19:14950033-15010643 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A6 | NM_001272087.1 | NP_001259016.1 | ||
| NM_001272088.1 | NP_001259017.1 | |||
| NM_005071.2 | NP_005062.1 | |||
| NR_073589.1 | ||||
| XM_006722842.1 | XP_006722905.1 | |||
| XM_006722844.1 | XP_006722907.1 | |||
| XM_017027152.1 | XP_016882641.1 | |||
| AK302380.1 | ||||
| AK312996.1 | ||||
| BC028721.2 | AAH28721.1 | |||
| BC040604.1 | AAH40604.1 | |||
| BI829415.1 | ||||
| U18244.1 | AAA75314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063744 | Chr.19:14906349 - 14999775 on Build GRCh38 | Gain |
|
| nsv2424 | Chr.19:14934808 - 14978815 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map