Assay Details
Target Gene Details
Entrez Gene ID: | 973 |
Gene Name: | CD79a molecule |
Gene Aliases: |
IGA, MB-1 |
Location: |
Chr.19:41877120-41881372 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD79A | NM_001783.3 | NP_001774.1 | ||
| NM_021601.3 | NP_067612.1 | |||
| AF283770.3 | AAG15590.1 | |||
| AK223371.1 | BAD97091.1 | |||
| AK315337.1 | ||||
| BC113731.1 | ||||
| BC113733.1 | ||||
| M74721.1 | AAA60270.1 | |||
| M80462.1 | AAA59556.1 | |||
| M86921.1 | AAA59557.1 | |||
| S46706.1 | AAB23558.1 | |||
| S75217.1 | AAB20812.1 | |||
| S79248.1 | AAC60653.1 | |||
| X83540.1 | CAA58523.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
| nsv953579 | Chr.19:41878331 - 41881433 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map