Assay Details
Target Gene Details
Entrez Gene ID: | 10054 |
Gene Name: | ubiquitin like modifier activating enzyme 2 |
Gene Aliases: |
ARX, HRIHFB2115, SAE2 |
Location: |
Chr.19:34428176-34469893 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA2 | NM_005499.2 | NP_005490.1 | ||
| XM_005258404.3 | XP_005258461.2 | |||
| XM_006722962.2 | XP_006723025.1 | |||
| XM_011526304.2 | XP_011524606.1 | |||
| XM_017026134.1 | XP_016881623.1 | |||
| AF079566.1 | AAD23914.1 | |||
| AF090384.1 | AAD12784.1 | |||
| AF110957.1 | AAD24434.1 | |||
| AK023120.1 | ||||
| AK024114.1 | ||||
| AK124730.1 | ||||
| AK299514.1 | ||||
| AK315521.1 | ||||
| AL136905.1 | CAB66839.1 | |||
| BC003153.1 | AAH03153.1 | |||
| BT009781.1 | AAP88783.1 | |||
| CR456756.1 | CAG33037.1 | |||
| U35832.1 | AAC99992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2469 | Chr.19:34442956 - 34476603 on Build GRCh38 | Insertion |
|
| nsv1059395 | Chr.19:34430094 - 34499592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map