Assay Details
Target Gene Details
Entrez Gene ID: | 100505622 |
Gene Name: | uncharacterized LOC100505622 |
Gene Aliases: |
- |
Location: |
Chr.19:42132629-42137099 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505622 | NR_038332.1 | 3 | 389 |
Target Gene Details
Entrez Gene ID: | 5452 |
Gene Name: | POU class 2 homeobox 2 |
Gene Aliases: |
OCT2, OTF2, Oct-2 |
Location: |
Chr.19:42086110-42197931 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU2F2 | XM_017026886.1 | XP_016882375.1 | ||
| XM_017026887.1 | XP_016882376.1 | |||
| XM_017026888.1 | XP_016882377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv2496 | Chr.19:42130710 - 42165604 on Build GRCh38 | Insertion |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map