Assay Details
Target Gene Details
Entrez Gene ID: | 10147 |
Gene Name: | SURP and G-patch domain containing 2 |
Gene Aliases: |
SFRS14 |
Location: |
Chr.19:18990887-19034052 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGP2 | NM_001017392.4 | NP_001017392.2 | ||
| NM_001321697.1 | NP_001308626.1 | |||
| NM_001321698.1 | NP_001308627.1 | |||
| NM_001321699.1 | NP_001308628.1 | |||
| NM_014884.4 | NP_055699.2 | |||
| XM_017026137.1 | XP_016881626.1 | |||
| AB002363.2 | ||||
| AF518874.1 | AAN77117.1 | |||
| AK074842.1 | ||||
| AK291275.1 | ||||
| AK299860.1 | ||||
| AL832488.1 | CAI46117.1 | |||
| BC020586.1 | AAH20586.1 | |||
| BX647813.1 | CAI46012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv2435 | Chr.19:18966623 - 19011770 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map