Assay Details
Target Gene Details
Entrez Gene ID: | 773 |
Gene Name: | calcium voltage-gated channel subunit alpha1 A |
Gene Aliases: |
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6 |
Location: |
Chr.19:13206442-13506460 on Build GRCh38 |
Assay Gene Location: | Within Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1A | NM_000068.3 | 48 | 8322 | NP_000059.3 |
| NM_001127221.1 | 47 | 8307 | NP_001120693.1 | |
| NM_001127222.1 | 47 | 8309 | NP_001120694.1 | |
| NM_001174080.1 | 48 | 8313 | NP_001167551.1 | |
| NM_023035.2 | 48 | 8327 | NP_075461.2 | |
| AF100774.1 | 19 | 3498 | AAC77460.1 | |
| EU016197.1 | 3 | 267 | ||
| U79668.1 | AAB49678.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs146613209] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map