Assay Details
Target Gene Details
Entrez Gene ID: | 23030 |
Gene Name: | lysine demethylase 4B |
Gene Aliases: |
JMJD2B, TDRD14B |
Location: |
Chr.19:4969112-5153603 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4B | NM_015015.2 | NP_055830.1 | ||
| XM_005259521.3 | XP_005259578.2 | |||
| XM_011527814.2 | XP_011526116.1 | |||
| XM_011527818.2 | XP_011526120.1 | |||
| XM_011527819.1 | XP_011526121.1 | |||
| XM_011527820.1 | XP_011526122.1 | |||
| XM_011527821.1 | XP_011526123.1 | |||
| XM_011527822.1 | XP_011526124.1 | |||
| XM_017026504.1 | XP_016881993.1 | |||
| XM_017026505.1 | XP_016881994.1 | |||
| AB020683.2 | ||||
| BC063889.1 | AAH63889.1 | |||
| DB636535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057373 | Chr.19:4963155 - 5037404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map