Assay Details
Target Gene Details
Entrez Gene ID: | 112703 |
Gene Name: | family with sequence similarity 71 member E1 |
Gene Aliases: |
- |
Location: |
Chr.19:50466643-50476746 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71E1 | NM_001308429.1 | NP_001295358.1 | ||
| NM_138411.2 | NP_612420.1 | |||
| XM_011526405.2 | XP_011524707.2 | |||
| XM_011526406.2 | XP_011524708.2 | |||
| AK125745.1 | ||||
| AK131032.1 | ||||
| AW665311.1 | ||||
| BC004941.1 | AAH04941.1 | |||
| BC012203.1 | AAH12203.1 | |||
| BC071737.1 | AAH71737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73579902] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map