Assay Details
Target Gene Details
Entrez Gene ID: | 9476 |
Gene Name: | napsin A aspartic peptidase |
Gene Aliases: |
KAP, Kdap, NAP1, NAPA, SNAPA |
Location: |
Chr.19:50358472-50369331 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPSA | NM_004851.2 | NP_004842.1 | ||
| XM_011527540.1 | 2 | 348 | XP_011525842.1 | |
| XM_017027512.1 | 2 | 348 | XP_016883001.1 | |
| BI770845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain |
|
| nsv2529 | Chr.19:50336652 - 50368507 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map