Assay Details
Target Gene Details
Entrez Gene ID: | 22900 |
Gene Name: | caspase recruitment domain family member 8 |
Gene Aliases: |
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN |
Location: |
Chr.19:48203148-48256269 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD8 | XM_017026485.1 | XP_016881974.1 | ||
| AF143869.1 | 4 | 356 | ||
| AK302327.1 | 10 | 1367 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
|
| nsv523716 | Chr.19:48198962 - 48208219 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map