Assay Details
Target Gene Details
Entrez Gene ID: | 25959 |
Gene Name: | KN motif and ankyrin repeat domains 2 |
Gene Aliases: |
ANKRD25, MXRA3, PPKWH, SIP |
Location: |
Chr.19:11164267-11197567 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK2 | NM_001136191.2 | NP_001129663.1 | ||
| NM_015493.6 | NP_056308.3 | |||
| AB040951.2 | ||||
| AB284125.1 | ||||
| AK002094.1 | BAA92081.1 | |||
| AK023332.1 | BAB14531.1 | |||
| AY639929.1 | AAT57879.1 | |||
| BC082762.1 | AAH82762.1 | |||
| BC098105.1 | AAH98105.1 | |||
| BC098286.1 | AAH98286.1 | |||
| BC098312.1 | AAH98312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3556028 | Chr.19:11184040 - 11185898 on Build GRCh38 | Deletion |
|
| nsv1065030 | Chr.19:11149372 - 11199804 on Build GRCh38 | Gain |
|
| esv2658280 | Chr.19:11184166 - 11185829 on Build GRCh38 | Deletion |
|
| esv3643666 | Chr.19:11184167 - 11185832 on Build GRCh38 | Loss |
|
| esv29212 | Chr.19:11184415 - 11185769 on Build GRCh38 | Loss |
|
| esv2718172 | Chr.19:11184151 - 11185799 on Build GRCh38 | Deletion |
|
| nsv578613 | Chr.19:11173352 - 11216950 on Build GRCh38 | Gain |
|
| esv3097124 | Chr.19:11184163 - 11185796 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map