Assay Details
Target Gene Details
Entrez Gene ID: | 23094 |
Gene Name: | signal induced proliferation associated 1 like 3 |
Gene Aliases: |
CTRCT45, SPAL3, SPAR3 |
Location: |
Chr.19:37906823-38208372 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIPA1L3 | NM_015073.2 | NP_055888.1 | ||
| XM_011526657.2 | XP_011524959.1 | |||
| XM_017026517.1 | XP_016882006.1 | |||
| XM_017026518.1 | XP_016882007.1 | |||
| BC036863.1 | AAH36863.1 | |||
| BC150620.1 | ||||
| BM193478.1 |
More Information
Set Membership: |
|

Genomic Map