Assay Details
Target Gene Details
Entrez Gene ID: | 55621 |
Gene Name: | tRNA methyltransferase 1 |
Gene Aliases: |
TRM1 |
Location: |
Chr.19:13104900-13117568 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT1 | NM_001136035.2 | NP_001129507.1 | ||
| NM_001142554.1 | NP_001136026.1 | |||
| NM_017722.3 | NP_060192.1 | |||
| XM_006722793.2 | XP_006722856.1 | |||
| XM_011528124.1 | XP_011526426.1 | |||
| XM_011528125.1 | XP_011526427.1 | |||
| XM_017026944.1 | XP_016882433.1 | |||
| XM_017026945.1 | XP_016882434.1 | |||
| XM_017026946.1 | XP_016882435.1 | |||
| XM_017026947.1 | XP_016882436.1 | |||
| AF196479.1 | AAG28495.1 | |||
| AK000251.1 | BAA91031.1 | |||
| AK292377.1 | ||||
| AK295245.1 | ||||
| AK304636.1 | ||||
| BC002492.2 | AAH02492.1 | |||
| BC018302.1 | AAH18302.1 | |||
| BC040126.1 | AAH40126.1 |
More Information
Set Membership: |
|

Genomic Map