Assay Details
Target Gene Details
Entrez Gene ID: | 1084 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 3 |
Gene Aliases: |
CD66D, CEA, CGM1, W264, W282 |
Location: |
Chr.19:41796590-41811553 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM3 | NM_001277163.2 | NP_001264092.1 | ||
| NM_001815.4 | NP_001806.2 | |||
| NR_102333.2 | ||||
| BC106728.1 | AAI06729.1 | |||
| D90277.1 | BAA14321.1 | |||
| D90278.1 | BAA14322.1 | |||
| E03348.1 | ||||
| E03349.1 | ||||
| L00692.1 | AAA51969.1 | |||
| L00693.1 | AAA51970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583420 | Chr.19:41802111 - 41814933 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
| esv3644395 | Chr.19:41806137 - 41816598 on Build GRCh38 | Loss |
|

Genomic Map