Assay Details
Target Gene Details
Entrez Gene ID: | 10523 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
DAN16, SCAF6, SRA1 |
Location: |
Chr.19:16517889-16542452 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHERP | NM_006387.5 | NP_006378.3 | ||
| AB209730.1 | BAD92967.1 | |||
| AF536542.1 | AAN77183.1 | |||
| AK296883.1 | ||||
| AK298408.1 | ||||
| BC021294.2 | AAH21294.1 | |||
| U94836.2 | AAB53327.1 |
More Information
Set Membership: |
|

Genomic Map