Assay Details
Target Gene Details
Entrez Gene ID: | 11202 |
Gene Name: | kallikrein related peptidase 8 |
Gene Aliases: |
HNP, NP, NRPN, PRSS19, TADG14 |
Location: |
Chr.19:50996008-51001702 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK8 | NM_001281431.1 | NP_001268360.1 | ||
| NM_007196.3 | NP_009127.1 | |||
| NM_144505.2 | NP_653088.1 | |||
| NM_144506.2 | NP_653089.1 | |||
| NM_144507.2 | NP_653090.1 | |||
| NR_104008.1 | ||||
| AA320217.1 | ||||
| AA587273.1 | ||||
| AB008390.2 | BAA82665.1 | |||
| AB008927.1 | BAA82666.1 | |||
| AB009849.1 | BAA28673.1 | |||
| AF095742.1 | AAD25979.1 | |||
| AY359036.1 | AAQ89395.1 | |||
| BM845575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644662 | Chr.19:50996542 - 51001686 on Build GRCh38 | Loss |
|
| nsv1058277 | Chr.19:50985288 - 51006591 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map