Assay Details
Target Gene Details
Entrez Gene ID: | 84658 |
Gene Name: | adhesion G protein-coupled receptor E3 |
Gene Aliases: |
EMR3 |
Location: |
Chr.19:14600175-14674918 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE3 | NM_001289158.1 | NP_001276087.1 | ||
| NM_001289159.1 | NP_001276088.1 | |||
| NM_032571.4 | NP_115960.2 | |||
| XM_011528374.2 | XP_011526676.1 | |||
| XM_017027383.1 | XP_016882872.1 | |||
| AF239764.1 | AAK15076.1 | |||
| AK290909.1 | ||||
| AK298653.1 | ||||
| AL832663.1 | ||||
| AY358817.1 | AAQ89176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953981 | Chr.19:14620089 - 14623388 on Build GRCh38 | Deletion |
|
| esv2534308 | Chr.19:14620893 - 14623781 on Build GRCh38 | Deletion |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
| nsv578665 | Chr.19:14620874 - 14623091 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67434392] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map