Assay Details
Target Gene Details
Entrez Gene ID: | 6597 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4 |
Gene Aliases: |
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b |
Location: |
Chr.19:10960922-11062282 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCA4 | NM_001128844.1 | NP_001122316.1 | ||
| NM_001128849.1 | NP_001122321.1 | |||
| NM_003072.3 | NP_003063.2 | |||
| XM_006722845.2 | XP_006722908.1 | |||
| XM_006722846.2 | XP_006722909.1 | |||
| XM_011528198.1 | XP_011526500.1 | |||
| XM_017027160.1 | XP_016882649.1 | |||
| XM_017027161.1 | XP_016882650.1 | |||
| XM_017027162.1 | XP_016882651.1 | |||
| XM_017027163.1 | XP_016882652.1 | |||
| XM_017027164.1 | XP_016882653.1 | |||
| XM_017027165.1 | XP_016882654.1 | |||
| XM_017027166.1 | XP_016882655.1 | |||
| XM_017027167.1 | XP_016882656.1 | |||
| XM_017027168.1 | XP_016882657.1 | |||
| AK290033.1 | ||||
| BC139917.1 | ||||
| BC139924.1 | ||||
| BC150298.1 | ||||
| BG396607.1 | ||||
| CT005139.1 | ||||
| D26156.1 | BAA05143.1 | |||
| U29175.1 | AAB40977.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map