Assay Details
Target Gene Details
Entrez Gene ID: | 51230 |
Gene Name: | PHD finger protein 20 |
Gene Aliases: |
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP |
Location: |
Chr.20:35771999-35950377 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF20 | NM_016436.4 | NP_057520.2 | ||
| XM_011528843.1 | XP_011527145.1 | |||
| XM_017027864.1 | XP_016883353.1 | |||
| XM_017027868.1 | XP_016883357.1 | |||
| XM_017027869.1 | XP_016883358.1 | |||
| XM_017027870.1 | XP_016883359.1 | |||
| XM_017027871.1 | XP_016883360.1 | |||
| AF220416.1 | AAF34184.1 | |||
| AF258787.1 | AAG49888.1 | |||
| AK054597.1 | ||||
| AK314503.1 | ||||
| AY134747.1 | AAN08621.1 | |||
| BC048210.1 | AAH48210.1 | |||
| BC080598.1 | AAH80598.1 | |||
| BC093405.1 | AAH93405.1 | |||
| BC150178.1 | ||||
| BG778940.1 | ||||
| BM906582.1 | ||||
| BX643875.1 | ||||
| CD690482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056503 | Chr.20:35750468 - 35805624 on Build GRCh38 | Gain |
|
| nsv526188 | Chr.20:35576541 - 36019079 on Build GRCh38 | Gain |
|
| dgv4303n100 | Chr.20:35730307 - 35805624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map