Assay Details
Target Gene Details
Entrez Gene ID: | 2139 |
Gene Name: | EYA transcriptional coactivator and phosphatase 2 |
Gene Aliases: |
EAB1 |
Location: |
Chr.20:46894624-47188844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA2 | NM_005244.4 | NP_005235.3 | ||
| NM_172110.3 | NP_742108.2 | |||
| XM_005260327.2 | XP_005260384.1 | |||
| XM_017027721.1 | XP_016883210.1 | |||
| AK293032.1 | ||||
| BC008803.2 | AAH08803.1 | |||
| BC013882.2 | AAH13882.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510788 | Chr.20:46884565 - 46990248 on Build GRCh38 | Deletion |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map