Assay Details
Target Gene Details
Entrez Gene ID: | 140885 |
Gene Name: | signal regulatory protein alpha |
Gene Aliases: |
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP |
Location: |
Chr.20:1894117-1939895 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPA | NM_001040022.1 | NP_001035111.1 | ||
| NM_001040023.1 | NP_001035112.1 | |||
| NM_080792.2 | NP_542970.1 | |||
| XM_005260669.3 | XP_005260726.1 | |||
| XM_005260670.3 | XP_005260727.1 | |||
| XM_006723545.3 | XP_006723608.1 | |||
| XM_011529173.2 | XP_011527475.1 | |||
| AB023430.1 | BAA87929.1 | |||
| AK290776.1 | ||||
| AK298243.1 | ||||
| AK312521.1 | ||||
| BC026692.1 | AAH26692.1 | |||
| BC033092.1 | AAH33092.1 | |||
| BC038510.2 | AAH38510.1 | |||
| BC075849.1 | AAH75849.1 | |||
| D86043.1 | BAA12974.1 | |||
| DN989886.1 | ||||
| Y10375.1 | CAA71403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1005896 | Chr.20:1914470 - 1915765 on Build GRCh38 | Gain |
|
| nsv511629 | Chr.20:1913689 - 1915094 on Build GRCh38 | Loss |
|
| dgv7437n54 | Chr.20:1914283 - 1915094 on Build GRCh38 | Loss |
|
| esv25632 | Chr.20:1914254 - 1915765 on Build GRCh38 | Gain+Loss |
|
| nsv585258 | Chr.20:1914433 - 1914987 on Build GRCh38 | Loss |
|
| nsv979411 | Chr.20:1913995 - 1916158 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs66675914,rs67414469] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map