Assay Details
Target Gene Details
Entrez Gene ID: | 80332 |
Gene Name: | ADAM metallopeptidase domain 33 |
Gene Aliases: |
C20orf153, DJ964F7.1 |
Location: |
Chr.20:3667973-3682131 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM33 | NM_001282447.2 | NP_001269376.1 | ||
| NM_025220.4 | NP_079496.1 | |||
| NM_153202.3 | NP_694882.1 | |||
| XM_005260843.1 | XP_005260900.1 | |||
| XM_006723639.1 | XP_006723702.1 | |||
| XM_006723640.1 | XP_006723703.1 | |||
| XM_011529366.1 | XP_011527668.1 | |||
| XM_011529367.1 | XP_011527669.1 | |||
| XM_011529368.1 | XP_011527670.1 | |||
| XM_011529373.1 | XP_011527675.1 | |||
| AB055891.1 | BAB83092.1 | |||
| AF466287.1 | AAM80482.1 | |||
| AK092627.1 | ||||
| AK094070.1 | 1 | 773 | ||
| AK095953.1 | 12 | 1809 | BAC04660.1 | |
| AK123015.1 | ||||
| AK126487.1 | ||||
| AK126498.1 | ||||
| AK226148.1 | ||||
| AL117415.1 | ||||
| AY358314.1 | AAQ88680.1 | |||
| BC125112.1 | ||||
| BC125113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575355 | Chr.20:3667953 - 3752840 on Build GRCh38 | Gain |
|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map