Assay Details
Target Gene Details
Entrez Gene ID: | 101927559 |
Gene Name: | CEBPB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:50184603-50192069 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPB-AS1 | NR_125739.1 | |||
| BX463443.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061982 | Chr.20:50111064 - 50483508 on Build GRCh38 | Loss |
|
| esv34129 | Chr.20:50132910 - 50304579 on Build GRCh38 | Loss |
|

Genomic Map