Assay Details
Target Gene Details
Entrez Gene ID: | 23394 |
Gene Name: | activity dependent neuroprotector homeobox |
Gene Aliases: |
ADNP1, HVDAS, MRD28 |
Location: |
Chr.20:50888918-50934984 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADNP | NM_001282531.1 | NP_001269460.1 | ||
| NM_001282532.1 | NP_001269461.1 | |||
| NM_015339.3 | NP_056154.1 | |||
| NM_181442.2 | NP_852107.1 | |||
| XM_011528747.2 | XP_011527049.1 | |||
| XM_011528748.2 | XP_011527050.1 | |||
| XM_017027757.1 | XP_016883246.1 | |||
| XM_017027758.1 | XP_016883247.1 | |||
| XM_017027759.1 | XP_016883248.1 | |||
| AB018327.1 | BAA34504.1 | |||
| AF250860.1 | AAG47651.1 | |||
| AK314734.1 | ||||
| BC075794.1 | AAH75794.1 | |||
| BC090933.1 | AAH90933.1 | |||
| DR006081.1 | ||||
| DR006382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834002 | Chr.20:50810235 - 50986764 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map