Assay Details
Target Gene Details
Entrez Gene ID: | 259240 |
Gene Name: | WAP four-disulfide core domain 9 |
Gene Aliases: |
WAP9, dJ688G8.2 |
Location: |
Chr.20:45607939-45631268 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC9 | NM_147198.3 | NP_671731.1 | ||
| AY047610.1 | AAK97772.1 | |||
| BC069295.1 | AAH69295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map