Assay Details
Target Gene Details
Entrez Gene ID: | 6789 |
Gene Name: | serine/threonine kinase 4 |
Gene Aliases: |
KRS2, MST1, YSK3 |
Location: |
Chr.20:44966474-45079977 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK4 | NM_006282.3 | NP_006273.1 | ||
| XM_005260530.2 | XP_005260587.1 | |||
| XM_005260531.3 | XP_005260588.1 | |||
| XM_005260532.3 | XP_005260589.1 | |||
| XM_005260533.2 | XP_005260590.1 | |||
| XM_011529018.2 | XP_011527320.1 | |||
| XM_011529020.2 | XP_011527322.1 | |||
| XM_017028029.1 | XP_016883518.1 | |||
| XM_017028030.1 | XP_016883519.1 | |||
| XM_017028031.1 | XP_016883520.1 | |||
| XM_017028032.1 | XP_016883521.1 | |||
| XM_017028033.1 | XP_016883522.1 | |||
| AB209315.1 | BAD92552.1 | |||
| AK309169.1 | ||||
| AK315238.1 | ||||
| AL597960.1 | ||||
| BC015332.1 | ||||
| BC029511.1 | AAH29511.1 | |||
| BC058916.1 | AAH58916.1 | |||
| BC093768.1 | AAH93768.1 | |||
| CR933633.1 | CAI45939.1 | |||
| U18297.1 | AAA83254.1 | |||
| U60207.1 | AAB17262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map