Assay Details
Target Gene Details
Entrez Gene ID: | 598 |
Gene Name: | BCL2 like 1 |
Gene Aliases: |
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52 |
Location: |
Chr.20:31664452-31723999 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L1 | NM_001191.3 | NP_001182.1 | ||
| NM_001317919.1 | NP_001304848.1 | |||
| NM_001317920.1 | NP_001304849.1 | |||
| NM_001317921.1 | NP_001304850.1 | |||
| NM_001322239.1 | NP_001309168.1 | |||
| NM_001322240.1 | NP_001309169.1 | |||
| NM_001322242.1 | NP_001309171.1 | |||
| NM_138578.2 | NP_612815.1 | |||
| NR_134257.1 | ||||
| XM_011528964.2 | XP_011527266.1 | |||
| XM_017027993.1 | XP_016883482.1 | |||
| AK290968.1 | ||||
| BC019307.1 | AAH19307.1 | |||
| BM554265.1 | ||||
| BT007208.1 | AAP35872.1 | |||
| BX359456.2 | ||||
| BX647525.1 | ||||
| CD359906.1 | ||||
| CR936637.1 | CAI56777.1 | |||
| DR156871.1 | ||||
| KU177938.1 | ||||
| KU177939.1 | ||||
| KU177940.1 | ||||
| Z23115.1 | CAA80661.1 | |||
| Z23116.1 | CAA80662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
| nsv585911 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111306603] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map