Assay Details
Target Gene Details
Entrez Gene ID: | 10816 |
Gene Name: | serine peptidase inhibitor, Kunitz type 3 |
Gene Aliases: |
HKIB9 |
Location: |
Chr.20:45512461-45515624 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINT3 | NM_006652.1 | 2 | 307 | NP_006643.1 |
| AY372172.2 | 2 | 307 | ||
| BX088813.1 | 2 | 257 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map